siRNA Details
| Gene Symbol: Tmem174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024298.1 | NP_001019469.1 | 1 | 582 | ||
| GenBank mRNA | BC079357.1 | - | 1 | 582 | 7108570 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem174| Gene Symbol: Tmem174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024298.1 | NP_001019469.1 | 1 | 582 | ||
| GenBank mRNA | BC079357.1 | - | 1 | 582 | 7108570 | |