siRNA Details
| Gene Symbol: Fam159b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574844.6 | XP_574844.3 | 2, 3 | 991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam159b| Gene Symbol: Fam159b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574844.6 | XP_574844.3 | 2, 3 | 991 | ||