siRNA Details
| Gene Symbol: Ndufs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025146.1 | NP_001020317.1 | 3 | 248 | ||
| GenBank mRNA | BC083729.1 | - | 3 | 248 | 7191329 | |
| BC127458.1 | - | 3 | 248 | 7193742 | ||
| FQ212751.1 | - | 3 | 265 | |||
| FQ214408.1 | - | 3 | 265 | |||
| FQ214557.1 | - | 3 | 265 | |||
| FQ223158.1 | - | 3 | 265 | |||

Ndufs4
Genomic Map