siRNA Details
| Gene Symbol: Lmbrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109177.1 | NP_001102647.1 | 11 | 1668 | ||
| XM_006232024.2 | XP_006232086.1 | 11 | 1659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmbrd2| Gene Symbol: Lmbrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109177.1 | NP_001102647.1 | 11 | 1668 | ||
| XM_006232024.2 | XP_006232086.1 | 11 | 1659 | |||