siRNA Details
| Gene Symbol: LOC499542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047943.1 | NP_001041408.1 | 2 | 104 | ||
| GenBank mRNA | AY539929.1 | - | 2 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499542| Gene Symbol: LOC499542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047943.1 | NP_001041408.1 | 2 | 104 | ||
| GenBank mRNA | AY539929.1 | - | 2 | 104 | ||