siRNA Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109178.1 | NP_001102648.1 | 5 | 825 | ||
| XM_008760792.1 | XP_008759014.1 | 4 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam173b| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109178.1 | NP_001102648.1 | 5 | 825 | ||
| XM_008760792.1 | XP_008759014.1 | 4 | 834 | |||