siRNA Details
| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024299.1 | NP_001019470.1 | 4 | 2169 | ||
| NM_001142962.1 | NP_001136434.1 | 4 | 2070 | |||
| XM_008760843.1 | XP_008759065.1 | 4, 5 | 2403 | |||
| GenBank mRNA | BC085784.1 | - | 4 | 2169 | 7107819 | |
| Gene Symbol: LOC100360380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760845.1 | XP_008759067.1 | 3 | 773 | ||

Zfp458
Genomic Map