siRNA Details
Gene Symbol: LOC499602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025039.2 | NP_001020210.2 | 13 | 1441 | ||
XM_006232310.2 | XP_006232372.1 | 12 | 1571 | |||
XM_006232311.2 | XP_006232373.1 | 9 | 1108 | |||
XM_008760940.1 | XP_008759162.1 | 12 | 1300 | |||
GenBank mRNA | BC086604.1 | - | 3 | 583 | 7317121 | |
BC098046.1 | - | 9 | 1080 |
