siRNA Details
| Gene Symbol: LOC499602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025039.2 | NP_001020210.2 | 4 | 332 | ||
| XM_006232310.2 | XP_006232372.1 | 4 | 552 | |||
| XM_008760940.1 | XP_008759162.1 | 3 | 191 | |||
| GenBank mRNA | CB612934.1 | - | 4 | 509 | ||
| CK358190.1 | - | 4 | 390 | |||

LOC499602
Genomic Map