siRNA Details
| Gene Symbol: Rfxap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044263.1 | NP_001037728.1 | 731 | |||
| GenBank mRNA | BC098816.1 | - | 731 | 7377196 | ||
| Gene Symbol: Rfxapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761021.1 | XP_008759243.1 | 3 | 758 | ||

Rfxap
Genomic Map