siRNA Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109184.1 | NP_001102654.1 | 5, 6 | 458 | ||
| XM_006232368.2 | XP_006232430.1 | 5, 6 | 2039 | |||
| XM_008761003.1 | XP_008759225.1 | 5, 6 | 2039 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc169| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109184.1 | NP_001102654.1 | 5, 6 | 458 | ||
| XM_006232368.2 | XP_006232430.1 | 5, 6 | 2039 | |||
| XM_008761003.1 | XP_008759225.1 | 5, 6 | 2039 | |||