siRNA Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109184.1 | NP_001102654.1 | 8 | 619 | ||
| XM_006232368.2 | XP_006232430.1 | 8 | 2200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc169| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109184.1 | NP_001102654.1 | 8 | 619 | ||
| XM_006232368.2 | XP_006232430.1 | 8 | 2200 | |||