siRNA Details
| Gene Symbol: RGD1565059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127562.1 | NP_001121034.1 | 2 | 228 | ||
| XM_006232447.2 | XP_006232509.1 | 2 | 188 | |||
| GenBank mRNA | BC167078.1 | - | 2 | 228 | 7133153 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565059| Gene Symbol: RGD1565059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127562.1 | NP_001121034.1 | 2 | 228 | ||
| XM_006232447.2 | XP_006232509.1 | 2 | 188 | |||
| GenBank mRNA | BC167078.1 | - | 2 | 228 | 7133153 | |