siRNA Details
Gene Symbol: RGD1565059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127562.1 | NP_001121034.1 | 3 | 346 | ||
XM_006232447.2 | XP_006232509.1 | 3 | 306 | |||
XM_006232448.2 | XP_006232510.1 | 2 | 246 | |||
XM_008761057.1 | XP_008759279.1 | 2 | 485 | |||
GenBank mRNA | BC167078.1 | - | 3 | 346 | 7133153 |
