siRNA Details
| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109185.1 | NP_001102655.1 | 5 | 647 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rit1| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109185.1 | NP_001102655.1 | 5 | 647 | ||