siRNA Details
| Gene Symbol: Gon4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024797.1 | NP_001019968.1 | 11 | 1732 | ||
| XM_006232735.2 | XP_006232797.1 | 11 | 1723 | |||
| XM_006232736.2 | XP_006232798.1 | 11 | 1708 | |||
| XM_006232737.2 | XP_006232799.1 | 11 | 1711 | |||
| XM_006232738.2 | XP_006232800.1 | 11 | 1708 | |||
| XM_006232739.2 | XP_006232801.1 | 11 | 1663 | |||
| XM_006232740.2 | XP_006232802.1 | 10 | 1519 | |||
| XM_006232741.2 | XP_006232803.1 | 11 | 1711 | |||
| XM_008761201.1 | XP_008759423.1 | 12 | 1737 | |||
| XM_008761202.1 | XP_008759424.1 | 10 | 1172 | |||
| GenBank mRNA | AB195689.1 | - | 10 | 1566 | ||
| AY724781.1 | - | 11 | 1732 | |||
| AY724782.1 | - | 11 | 1732 | |||

Gon4l
Genomic Map