siRNA Details
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024797.1 | NP_001019968.1 | 12 | 1930 | ||
XM_006232735.2 | XP_006232797.1 | 12 | 1921 | |||
XM_006232736.2 | XP_006232798.1 | 12 | 1906 | |||
XM_006232737.2 | XP_006232799.1 | 12 | 1909 | |||
XM_006232738.2 | XP_006232800.1 | 12 | 1906 | |||
XM_006232739.2 | XP_006232801.1 | 12 | 1861 | |||
XM_006232740.2 | XP_006232802.1 | 11 | 1717 | |||
XM_006232741.2 | XP_006232803.1 | 12 | 1909 | |||
XM_008761201.1 | XP_008759423.1 | 13 | 1935 | |||
XM_008761202.1 | XP_008759424.1 | 11 | 1370 | |||
GenBank mRNA | AB195689.1 | - | 11 | 1764 | ||
AY724781.1 | - | 12 | 1930 | |||
AY724782.1 | - | 12 | 1930 |
