siRNA Details
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024797.1 | NP_001019968.1 | 31 | 6308 | ||
XM_006232735.2 | XP_006232797.1 | 31 | 6299 | |||
XM_006232736.2 | XP_006232798.1 | 31 | 6284 | |||
XM_006232737.2 | XP_006232799.1 | 30 | 6263 | |||
XM_006232738.2 | XP_006232800.1 | 30 | 6260 | |||
XM_006232739.2 | XP_006232801.1 | 31 | 6239 | |||
XM_006232740.2 | XP_006232802.1 | 30 | 6095 | |||
XM_006232741.2 | XP_006232803.1 | 30 | 6089 | |||
XM_008761201.1 | XP_008759423.1 | 32 | 6313 | |||
XM_008761202.1 | XP_008759424.1 | 30 | 5748 | |||
GenBank mRNA | AB195689.1 | - | 30 | 6142 | ||
AY724781.1 | - | 31 | 6308 | |||
AY724782.1 | - | 30 | 6388 |
