siRNA Details
| Gene Symbol: RGD1564171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109186.1 | NP_001102656.1 | 4 | 316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564171| Gene Symbol: RGD1564171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109186.1 | NP_001102656.1 | 4 | 316 | ||