siRNA Details
| Gene Symbol: Celf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109190.2 | NP_001102660.1 | 2, 3 | 985 | ||
| XM_006232897.2 | XP_006232959.1 | 2, 3 | 911 | |||
| XM_006232898.2 | XP_006232960.1 | 2, 3 | 912 | |||
| XM_006232900.2 | XP_006232962.1 | 2, 3 | 911 | |||
| XM_006232902.2 | XP_006232964.1 | 2, 3 | 911 | |||
| XM_006232904.2 | XP_006232966.1 | 3, 4 | 2639 | |||
| XM_006232905.2 | XP_006232967.1 | 3, 4 | 2672 | |||
| XM_006232908.2 | XP_006232970.1 | 2, 3 | 909 | |||
| XM_008761322.1 | XP_008759544.1 | 3, 4 | 3226 | |||
| XM_008761323.1 | XP_008759545.1 | 2, 3 | 195 | |||

Celf3
Genomic Map