siRNA Details
| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039044.1 | NP_001034133.1 | 4 | 494 | ||
| GenBank mRNA | BC091418.1 | - | 4 | 494 | 7365287 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc42se1| Gene Symbol: Cdc42se1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039044.1 | NP_001034133.1 | 4 | 494 | ||
| GenBank mRNA | BC091418.1 | - | 4 | 494 | 7365287 | |