siRNA Details
Gene Symbol: Gstm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109192.1 | NP_001102662.1 | 1 | 125 | ||
GenBank mRNA | BC158681.1 | - | 1 | 135 | 8373383 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gstm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109192.1 | NP_001102662.1 | 1 | 125 | ||
GenBank mRNA | BC158681.1 | - | 1 | 135 | 8373383 |