siRNA Details
| Gene Symbol: Gstm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109192.1 | NP_001102662.1 | 6 | 494 | ||
| GenBank mRNA | BC158681.1 | - | 6 | 504 | 8373383 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstm6| Gene Symbol: Gstm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109192.1 | NP_001102662.1 | 6 | 494 | ||
| GenBank mRNA | BC158681.1 | - | 6 | 504 | 8373383 | |