siRNA Details
| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100993.1 | NP_001094463.1 | 7 | 966 | ||
| XM_008761520.1 | XP_008759742.1 | 7 | 1060 | |||
| GenBank mRNA | BC128777.1 | - | 7 | 966 | 7134244 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stpg2| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100993.1 | NP_001094463.1 | 7 | 966 | ||
| XM_008761520.1 | XP_008759742.1 | 7 | 1060 | |||
| GenBank mRNA | BC128777.1 | - | 7 | 966 | 7134244 | |