siRNA Details
Gene Symbol: RGD1560065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134616.1 | NP_001128088.1 | 2 | 302 | ||
XM_006233461.2 | XP_006233523.1 | 2 | 381 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1560065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134616.1 | NP_001128088.1 | 2 | 302 | ||
XM_006233461.2 | XP_006233523.1 | 2 | 381 |