siRNA Details
| Gene Symbol: Il36g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113790.1 | NP_001107262.1 | 5 | 572 | ||
| XM_008761605.1 | XP_008759827.1 | 5 | 565 | |||
| GenBank mRNA | BC158611.1 | - | 5 | 572 | 7384725 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il36g| Gene Symbol: Il36g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113790.1 | NP_001107262.1 | 5 | 572 | ||
| XM_008761605.1 | XP_008759827.1 | 5 | 565 | |||
| GenBank mRNA | BC158611.1 | - | 5 | 572 | 7384725 | |