siRNA Details
| Gene Symbol: Rnf208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109195.1 | NP_001102665.1 | 2 | 880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf208| Gene Symbol: Rnf208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109195.1 | NP_001102665.1 | 2 | 880 | ||