siRNA Details
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024309.2 | NP_001019480.2 | 1 | 1626 | ||
GenBank mRNA | BC083928.1 | - | 1 | 960 | 7135551 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024309.2 | NP_001019480.2 | 1 | 1626 | ||
GenBank mRNA | BC083928.1 | - | 1 | 960 | 7135551 |