siRNA Details
| Gene Symbol: RGD1564379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126295.1 | NP_001119767.1 | 2 | 726 | ||
| XM_006233690.2 | XP_006233752.1 | 2 | 798 | |||
| XM_006233691.2 | XP_006233753.1 | 2 | 726 | |||
| XM_006233692.1 | XP_006233754.1 | 2 | 552 | |||
| GenBank mRNA | BC161868.1 | - | 2 | 726 | 7108986 | |

RGD1564379
Genomic Map