siRNA Details
| Gene Symbol: RGD1564114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127563.1 | NP_001121035.1 | 6 | 858 | ||
| GenBank mRNA | BC166521.1 | - | 6 | 858 | 7133997 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564114| Gene Symbol: RGD1564114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127563.1 | NP_001121035.1 | 6 | 858 | ||
| GenBank mRNA | BC166521.1 | - | 6 | 858 | 7133997 | |