siRNA Details
| Gene Symbol: LOC499770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047110.1 | NP_001040575.1 | 5, 6 | 540 | ||
| XM_006233840.2 | XP_006233902.1 | 8, 9 | 1214 | |||
| XM_006233841.2 | XP_006233903.1 | 8, 9 | 1003 | |||
| XM_006233842.2 | XP_006233904.1 | 8, 9 | 791 | |||
| XM_008761672.1 | XP_008759894.1 | 9, 10 | 867 | |||
| GenBank mRNA | BC098818.1 | - | 5, 6 | 540 | 7377299 | |

LOC499770
Genomic Map