siRNA Details
| Gene Symbol: Ier5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025041.1 | NP_001020212.1 | 1 | 1110 | ||
| GenBank mRNA | BC087095.1 | - | 1 | 1110 | 7191438 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ier5l| Gene Symbol: Ier5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025041.1 | NP_001020212.1 | 1 | 1110 | ||
| GenBank mRNA | BC087095.1 | - | 1 | 1110 | 7191438 | |