siRNA Details
| Gene Symbol: LOC499796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047951.1 | NP_001041416.1 | 5 | 607 | ||
| GenBank mRNA | AY383667.1 | - | 5 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499796| Gene Symbol: LOC499796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047951.1 | NP_001041416.1 | 5 | 607 | ||
| GenBank mRNA | AY383667.1 | - | 5 | 607 | ||