siRNA Details
| Gene Symbol: Arl6ip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024310.1 | NP_001019481.1 | 1 | 379 | ||
| GenBank mRNA | BC079329.1 | - | 1 | 379 | 7096035 | |
| FQ222449.1 | - | 385 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl6ip6| Gene Symbol: Arl6ip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024310.1 | NP_001019481.1 | 1 | 379 | ||
| GenBank mRNA | BC079329.1 | - | 1 | 379 | 7096035 | |
| FQ222449.1 | - | 385 | ||||