siRNA Details
| Gene Symbol: Ifih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109199.1 | NP_001102669.1 | 5 | 1304 | ||
| GenBank mRNA | BC168680.1 | - | 2 | 329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifih1| Gene Symbol: Ifih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109199.1 | NP_001102669.1 | 5 | 1304 | ||
| GenBank mRNA | BC168680.1 | - | 2 | 329 | ||