siRNA Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024311.1 | NP_001019482.1 | 9 | 1167 | ||
| XM_006234340.2 | XP_006234402.1 | 9 | 2273 | |||
| XM_008761921.1 | XP_008760143.1 | 2 | 318 | |||
| GenBank mRNA | BC082109.1 | - | 9 | 1167 | 7135070 | |

Erich2
Genomic Map