siRNA Details
| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177688.1 | NP_001171159.1 | 9 | 1078 | ||
| XM_006234347.2 | XP_006234409.1 | 9 | 1150 | |||
| GenBank mRNA | BC160919.1 | - | 9 | 1078 | 7458373 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcaf17| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177688.1 | NP_001171159.1 | 9 | 1078 | ||
| XM_006234347.2 | XP_006234409.1 | 9 | 1150 | |||
| GenBank mRNA | BC160919.1 | - | 9 | 1078 | 7458373 | |