siRNA Details
| Gene Symbol: LOC499843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047953.1 | NP_001041418.1 | 3 | 123 | ||
| GenBank mRNA | AY387077.1 | - | 3 | 123 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499843| Gene Symbol: LOC499843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047953.1 | NP_001041418.1 | 3 | 123 | ||
| GenBank mRNA | AY387077.1 | - | 3 | 123 | ||