siRNA Details
| Gene Symbol: Rad51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109204.1 | NP_001102674.1 | 4 | 510 | ||
| XM_006234784.2 | XP_006234846.1 | 5 | 688 | |||
| GenBank mRNA | BC168875.1 | - | 4 | 474 | 9022295 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad51| Gene Symbol: Rad51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109204.1 | NP_001102674.1 | 4 | 510 | ||
| XM_006234784.2 | XP_006234846.1 | 5 | 688 | |||
| GenBank mRNA | BC168875.1 | - | 4 | 474 | 9022295 | |