siRNA Details
| Gene Symbol: Rad51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109204.1 | NP_001102674.1 | 8 | 947 | ||
| XM_006234784.2 | XP_006234846.1 | 9 | 1125 | |||
| GenBank mRNA | BC168875.1 | - | 8 | 911 | 9022295 | |
| Gene Symbol: LOC100911267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762135.1 | XP_008760357.1 | 4 | 484 | ||

Rad51
Genomic Map