siRNA Details
| Gene Symbol: Eid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109205.1 | NP_001102675.1 | 1 | 306 | ||
| GenBank mRNA | BC162030.1 | - | 1 | 242 | 9100917 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eid1| Gene Symbol: Eid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109205.1 | NP_001102675.1 | 1 | 306 | ||
| GenBank mRNA | BC162030.1 | - | 1 | 242 | 9100917 | |