siRNA Details
Gene Symbol: Zfp133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024313.1 | NP_001019484.2 | 4 | 736 | ||
GenBank mRNA | BC079070.1 | - | 4 | 736 | 7121746 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024313.1 | NP_001019484.2 | 4 | 736 | ||
GenBank mRNA | BC079070.1 | - | 4 | 736 | 7121746 |