siRNA Details
| Gene Symbol: Napb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191966.1 | NP_001178895.1 | 11 | 834 | ||
| XM_008762275.1 | XP_008760497.1 | 9 | 689 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Napb| Gene Symbol: Napb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191966.1 | NP_001178895.1 | 11 | 834 | ||
| XM_008762275.1 | XP_008760497.1 | 9 | 689 | |||