siRNA Details
| Gene Symbol: Bpifb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077680.1 | NP_001071148.2 | 9 | 821 | ||
| XM_006235355.2 | XP_006235417.1 | 9 | 900 | |||
| GenBank mRNA | BC127515.1 | - | 9 | 821 | 7100079 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifb1| Gene Symbol: Bpifb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077680.1 | NP_001071148.2 | 9 | 821 | ||
| XM_006235355.2 | XP_006235417.1 | 9 | 900 | |||
| GenBank mRNA | BC127515.1 | - | 9 | 821 | 7100079 | |