siRNA Details
| Gene Symbol: RGD1561517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135901.1 | NP_001129373.1 | 3 | 542 | ||
| XM_006235356.2 | XP_006235418.1 | 3 | 1950 | |||
| XM_006235357.2 | XP_006235419.1 | 2 | 1853 | |||
| GenBank mRNA | DQ080432.1 | - | 3 | 542 | ||

RGD1561517
Genomic Map