siRNA Details
| Gene Symbol: Dsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025768.1 | NP_001020939.1 | 3 | 122 | ||
| XM_006235375.2 | XP_006235437.1 | 3 | 432 | |||
| XM_006235376.2 | XP_006235438.1 | 3 | 1103 | |||
| XM_006235377.2 | XP_006235439.1 | 3 | 1195 | |||
| XM_006235378.2 | XP_006235440.1 | 3 | 432 | |||
| GenBank mRNA | BC098027.1 | - | 3 | 122 | 7456020 | |

Dsn1
Genomic Map