siRNA Details
| Gene Symbol: RGD1564340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134620.1 | NP_001128092.1 | 2 | 625 | ||
| XM_006235792.2 | XP_006235854.1 | 2 | 1570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564340| Gene Symbol: RGD1564340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134620.1 | NP_001128092.1 | 2 | 625 | ||
| XM_006235792.2 | XP_006235854.1 | 2 | 1570 | |||