siRNA Details
| Gene Symbol: Samd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191967.1 | NP_001178896.1 | 4 | 459 | ||
| XM_006235798.1 | XP_006235860.1 | 4 | 470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd10| Gene Symbol: Samd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191967.1 | NP_001178896.1 | 4 | 459 | ||
| XM_006235798.1 | XP_006235860.1 | 4 | 470 | |||