siRNA Details
| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025148.1 | NP_001020319.1 | 2, 3 | 330 | ||
| GenBank mRNA | BC078995.1 | - | 2, 3 | 330 | 7112721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galntl5| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025148.1 | NP_001020319.1 | 2, 3 | 330 | ||
| GenBank mRNA | BC078995.1 | - | 2, 3 | 330 | 7112721 | |