siRNA Details
| Gene Symbol: LOC499971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024317.2 | NP_001019488.2 | 9, 10 | 1229 | ||
| XM_008762655.1 | XP_008760877.1 | 8, 9 | 882 | |||
| XM_008762656.1 | XP_008760878.1 | 7, 8 | 773 | |||
| GenBank mRNA | BC079392.1 | - | 2, 3 | 165 | 7111961 | |

LOC499971
Genomic Map